missing translation for 'onlineSavingsMsg'
Learn More

(4R,6R)-tert-Butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 97%, Thermo Scientific™

Catalog No. AC357990010
Change view
Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC357990010 1g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC357990010 Supplier Thermo Scientific Chemicals Supplier No. 357990010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 125971-94-0
Molecular Formula C14H23NO4
Molecular Weight (g/mol) 269.34
MDL Number MFCD03093958
InChI Key DPNRMEJBKMQHMC-GHMZBOCLSA-N
Synonym 4r,6r-tert-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 4r,6r-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 2-4r,6r-6-cyanomethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate, 4r,6r-t-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r-cis-1,1-dimethylethyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, pubchem5924, 3r,5r-6-cyano-3,5-isopropylidenebisoxy hexanoic acid tert-butyl ester, 4r,3r-tert-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,6r-tert-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4acetate
PubChem CID 2734287
IUPAC Name tert-butyl 2-[(4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMILES CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC#N)OC(C)(C)O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1g
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform
Formula Weight 269.34
Percent Purity 97%
Chemical Name or Material (4R,6R)-tert-Butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 97%

RUO – Research Use Only