accessibility menu, dialog, popup

UserName

H3COH3CCH3HNNNNO

CAS RN 1256580-46-7

IUPAC Name: 9-ethyl-6,6-dimethyl-8-[4-(morpholin-4-yl)piperidin-1-yl]-11-oxo-5H,6H,11H-benzo[b]carbazole-3-carbonitrile
Synonyms: alectinib
Molecular Weight (g/mol): 482.63
Molecular Formula: C30H34N4O2
InChi Key: KDGFLJKFZUIJMX-UHFFFAOYSA-N
SMILES: CCC1=C(C=C2C(=C1)C(=O)C1=C(NC3=CC(=CC=C13)C#N)C2(C)C)N1CCC(CC1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)