accessibility menu, dialog, popup

UserName

H3CCH3HOHOOOHCH3H3CH3CCH3OHOHOOHCH3OHO(?)

CAS RN 12542-36-8

IUPAC Name: 1,1',6,6',7,7'-hexahydroxy-3,3'-dimethyl-5,5'-bis(propan-2-yl)-[2,2'-binaphthalene]-8,8'-dicarbaldehyde; acetic acid
Synonyms: (-)-gossypol; acetic acid
Molecular Weight (g/mol): 578.61
Molecular Formula: C32H34O10
InChi Key: NIOHNDKHQHVLKA-UHFFFAOYSA-N
SMILES: CC(O)=O.CC(C)C1=C(O)C(O)=C(C=O)C2=C(O)C(=C(C)C=C12)C1=C(O)C2=C(C=O)C(O)=C(O)C(C(C)C)=C2C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results