missing translation for 'onlineSavingsMsg'
Learn More

Gossypol-acetic acid

Catalog No. AAJ63517MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63517MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63517MC Supplier Thermo Scientific Chemicals Supplier No. J63517MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12542-36-8 | C32H34O10 | 578.614 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12542-36-8
Molecular Formula C32H34O10
Molecular Weight (g/mol) 578.614
MDL Number MFCD00058385
InChI Key NIOHNDKHQHVLKA-UHFFFAOYSA-N
Synonym gossypol-acetic acid, gossypol acetate, gossypol acetic acid, acetate gossypol, --gossypol acetic acid, s-gossypol acetic acid, at-101 acetic acid, gossypol-acetic acid complex
PubChem CID 227456
IUPAC Name acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
SMILES CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O

Specifications

Color Yellow to Green
Quantity 100 mg
Solubility Information Soluble in acetone or methanol at 5mg/ml
Formula Weight 578.61
Physical Form Powder
Chemical Name or Material Gossypol-acetic acid
Hazard Category H302-H351
Hazard Statement GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
RTECSNumber MD7360000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only