accessibility menu, dialog, popup

UserName

NH2NH2OOHHO

CAS RN 124-43-6

IUPAC Name: peroxol; urea
Synonyms: hydrogen peroxide - urea
Molecular Weight (g/mol): 94.07
Molecular Formula: CH6N2O3
InChi Key: AQLJVWUFPCUVLO-UHFFFAOYSA-N
SMILES: OO.NC(N)=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Grade 
  • (9)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (15)
  • (9)
  • (9)
  • (6)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (9)
  • (3)
  • (3)
  • (3)