missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals

Catalog No. AC163670010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163670010 1 L Glass bottle
AC163670025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163670010 Supplier Thermo Scientific Chemicals Supplier No. 163670010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Dimethylamine
CAS 124-40-3, 7732-18-5
Melting Point -37.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 54.0°C
Flash Point −18°C
Packaging Glass bottle
Molecular Formula C2H7N
Refractive Index 1.37
Linear Formula (CH3)2NH
Quantity 1 L
MDL Number MFCD00008288
Beilstein 04, 39
Fieser 07,119
Specific Gravity 0.89
Merck Index 15, 3250
Synonym dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in alcohol and ether
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES CNC
IUPAC Name N-methylmethanamine
Molecular Weight (g/mol) 45.07
PubChem CID 674
ChEBI CHEBI:17170
Formula Weight 45.07
Percent Purity ≥40%
Physical Form Liquid
Show More Show Less

RUO – Research Use Only