missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals

Catalog No. AA43261KW
Change view
Click to view available options
Quantity:
10 Ea.
50 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43261KW 50 Ea.
AA43261KV 10 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43261KW Supplier Thermo Scientific Chemicals Supplier No. 043261KW

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dimethylamine is a used as a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber.

Dimethylamine is a used as a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrochemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-40-3
Molecular Formula C2H7N
Molecular Weight (g/mol) 45.085
MDL Number MFCD00008288
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
Synonym dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
PubChem CID 674
ChEBI CHEBI:17170
IUPAC Name N-methylmethanamine
SMILES CNC

Specifications

Density 0.89
Flash Point 15°C (60°F)
Packaging Ampoule
Refractive Index 1.37
Quantity 50 Ea.
UN Number UN1160
Beilstein 605257
Specific Gravity 0.89
Solubility Information Fully miscible in water.
Formula Weight 45.08
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dimethylamine
Show More Show Less
Hazard Category H225-H302+H332-H314-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 204-697-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only