CAS RN 1232410-49-9
IUPAC Name:
3-amino-6-(4-methanesulfonylphenyl)-N-phenylpyrazine-2-carboxamide
Synonyms:
3-amino-6-(4-methanesulfonylphenyl)-N-phenylpyrazine-2-carboxamide
Molecular Weight (g/mol):
368.41
Molecular Formula:
C18H16N4O3S
InChi Key:
DUIHHZKTCSNTGM-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C1=CC=C(C=C1)C1=CN=C(N)C(=N1)C(=O)NC1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals VE-821, 95%
CAS: 1232410-49-9 Molecular Formula: C18H16N4O3S