accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 123-66-0

IUPAC Name: ethyl hexanoate
Synonyms: ethyl hexanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: SHZIWNPUGXLXDT-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OCC
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)