missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hexanoate, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019518
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019518 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019518 Supplier Merck Emd Millipore Supplier No. 083751ML

Specifications

CAS 123-66-0
Vapor Density 5 (vs. air)
Density 0.869 g/mL (at 25°C)
Linear Formula CH3(CH2)4 COOC2H5
MDL Number MFCD00009511
Boiling Point 168°C
Synonym Caproic acid ethyl ester; Ethyl caproate
Quantity 1 mL
Formula Weight 144.21
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less