accessibility menu, dialog, popup

UserName

CH3OOCH3

CAS RN 121-97-1

IUPAC Name: 1-(4-methoxyphenyl)propan-1-one
Synonyms: P-methoxypropiophenone
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: ZJVAWPKTWVFKHG-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(OC)C=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)