CAS RN 12055-62-8
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh
CAS: 12055-62-8 Molecular Formula: Ho2O3 Molecular Weight (g/mol): 377.86 MDL Number: MFCD00011053 InChI Key: OWCYYNSBGXMRQN-UHFFFAOYSA-N IUPAC Name: diholmium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Ho+3].[Ho+3]
Holmium(III) oxide, REacton™, 99.9% (REO)
CAS: 12055-62-8 Molecular Formula: Ho2O3 Molecular Weight (g/mol): 377.86 MDL Number: MFCD00011053 InChI Key: OWCYYNSBGXMRQN-UHFFFAOYSA-N IUPAC Name: diholmium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Ho+3].[Ho+3]
Holmium ICP Standard, 1000 ppm Ho in 3% HNO3, Ricca Chemical
CAS: 12055-62-8
Holmium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 12055-62-8 Molecular Formula: Ho2O3 Molecular Weight (g/mol): 377.86 MDL Number: MFCD00011053 InChI Key: OWCYYNSBGXMRQN-UHFFFAOYSA-N IUPAC Name: diholmium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Ho+3].[Ho+3]
Holmium Standard For ICP, 1000μg/mL in 2% HNO3, SPEX CertiPrep™
CAS: 12055-62-8 Molecular Formula: Ho2O3 Molecular Weight (g/mol): 377.86 MDL Number: MFCD00011053 InChI Key: OWCYYNSBGXMRQN-UHFFFAOYSA-N IUPAC Name: diholmium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Ho+3].[Ho+3]
Holmium Standard For ICP, 1000μg/mL, SPEX CertiPrep™
CAS: 12055-62-8 Molecular Formula: Ho2O3 Molecular Weight (g/mol): 377.86 MDL Number: MFCD00011053 InChI Key: OWCYYNSBGXMRQN-UHFFFAOYSA-N IUPAC Name: diholmium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Ho+3].[Ho+3]