accessibility menu, dialog, popup

UserName

CAS RN 120511-73-1

IUPAC Name: 2-[3-(1-cyano-1-methylethyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]phenyl]-2-methylpropanenitrile
Synonyms: anastrozole zeneca zd 1033 阿那曲唑 anastrozole (product) zeneca ZD 1033 astrazeneca brand of anastrozole anastrole anastrazole zeneca brand of anastrozole anastrozole (substance)
Molecular Weight (g/mol): 293.37
Molecular Formula: C17H19N5
InChi Key: YBBLVLTVTVSKRW-UHFFFAOYSA-N
SMILES: CC(C)(C#N)C1=CC(=CC(CN2C=NC=N2)=C1)C(C)(C)C#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)