accessibility menu, dialog, popup

UserName

CAS RN 1205-02-3

CH3NHOOHO

CAS RN 1205-02-3

IUPAC Name: 2-(phenylformamido)propanoic acid
Synonyms: methylhippuric acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: UAQVHNZEONHPQG-UHFFFAOYNA-N
SMILES: CC(NC(=O)C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)