missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl-DL-alanine 98.0+%, TCI America™

Catalog No. B013525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B013525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B013525G Supplier TCI America Supplier No. B013525G

Chemical Identifiers

CAS 1205-02-3
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00020393
InChI Key UAQVHNZEONHPQG-UHFFFAOYNA-N
Synonym n-benzoyl-dl-alanine, benzoyl-dl-alanine, methylhippuric acid, n-benzoylalanine, alanine, n-benzoyl, benzoylalanine, dl-n-benzoylalanine, 2-phenylformamido propanoic acid, alpha-methylhippuric acid, n-phenylcarbonyl alanine
PubChem CID 71002
ChEBI CHEBI:71167
IUPAC Name 2-(phenylformamido)propanoic acid
SMILES CC(NC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 166°C
Color White
Quantity 25 g
Formula Weight 193.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl-DL-alanine