accessibility menu, dialog, popup

UserName

H3COO(S)NSClOHSOHOO

CAS RN 120202-66-6

IUPAC Name: methyl (2S)-2-(2-chlorophenyl)-2-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}acetate; sulfuric acid
Synonyms: clopidogrel; sulfuric acid
Molecular Weight (g/mol): 419.89
Molecular Formula: C16H18ClNO6S2
InChi Key: FDEODCTUSIWGLK-RSAXXLAASA-N
SMILES: OS(O)(=O)=O.COC(=O)[C@@H](N1CCC2=C(C1)C=CS2)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)