missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clopidogrel Bisulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Clopidogrel Bisulfate is a novel antiplatelet agent which is chemically related to ticlopidine. It acts against ischaemic stroke, myocardial infraction as well as vascular disease. It is reported to have exhibited clinical efficacy greater than that of aspirin, during large phase III trials. Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 120202-66-6 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C16H16 ClNO2 S · H2 SO4 |
| MDL Number | MFCD00876395 |
| Synonym | -Methyl (2-chlorophenyl)(6,7-dihydro-4 H-thieno[3,2-c]pyridin-5-yl)acetate hydrogensulfate |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 419.90 |
| Grade | Certified Reference Material |