accessibility menu, dialog, popup

UserName

H3COOCH3H3CO

CAS RN 1201-38-3

IUPAC Name: 1-(2,5-dimethoxyphenyl)ethan-1-one
Synonyms: 1-(2,5-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: FAXUIYJKGGUCBO-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(=C1)C(C)=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)