accessibility menu, dialog, popup

UserName

CH3NOH

CAS RN 120-29-6

IUPAC Name: 8-methyl-8-azabicyclo[3.2.1]octan-3-ol
Synonyms: tropin
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: CYHOMWAPJJPNMW-UHFFFAOYNA-N
SMILES: CN1C2CCC1CC(O)C2
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)