missing translation for 'onlineSavingsMsg'
Learn More

Tropine, 98%

Catalog No. AAL0233218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0233218 50 g
AAL0233209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0233218 Supplier Thermo Scientific Chemicals Supplier No. L0233218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120-29-6 | C8H15NO | 141.214 g/mol

Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

Solubility
Soluble in water. (100 g/L) at 20°C.

Notes
Hygroscopic. Store at 4°C. Incomaptible with Strong acids and oxidizing agents.

Chemical Identifiers

CAS 120-29-6
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00005551
InChI Key CYHOMWAPJJPNMW-RNFRBKRXSA-N
Synonym tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
PubChem CID 6101956
IUPAC Name (1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
SMILES CN1C2CCC1CC(C2)O

Specifications

Melting Point 61°C to 65°C (dried)
Density 1.04
pH 12
Boiling Point 232°C to 234°C
Odor Amine-like
Quantity 50 g
Beilstein 80188
Sensitivity Hygroscopic
Merck Index 14,9781
Solubility Information Soluble in water. (100g/L) at 20°C.
Formula Weight 141.22
Percent Purity 98%
Chemical Name or Material Tropine
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 204-384-2
RTECSNumber YM3875000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only