accessibility menu, dialog, popup

UserName

NH2NOClHN

CAS RN 1194-21-4

IUPAC Name: 2-amino-6-chloro-1,4-dihydropyrimidin-4-one
Synonyms: 2-amino-6-chloro-1H-pyrimidin-4-one
Molecular Weight (g/mol): 145.55
Molecular Formula: C4H4ClN3O
InChi Key: VBWACOJLJYUFKJ-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C=C(Cl)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results