accessibility menu, dialog, popup

UserName

H3COO

CAS RN 1192-62-7

IUPAC Name: 1-(furan-2-yl)ethan-1-one
Synonyms: acetylfuran (2-furanyl)-1-ethanone 2-furylethanone furyl methyl ketone
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: IEMMBWWQXVXBEU-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CO1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)