missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylfuran, 99%

Catalog No. AC102552500
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102552500 250 g Glass bottle
AC102551000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102552500 Supplier Thermo Scientific Chemicals Supplier No. 102552500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1192-62-7 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1192-62-7
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00003242
InChI Key IEMMBWWQXVXBEU-UHFFFAOYSA-N
Synonym 2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
PubChem CID 14505
ChEBI CHEBI:59983
IUPAC Name 1-(furan-2-yl)ethanone
SMILES CC(=O)C1=CC=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29.0°C to 30.0°C
Color Yellow to Brown
Density 1.0980g/mL
Boiling Point 67.0°C (10.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. 2,5-Diacetyl furan (GC)
Packaging Glass bottle
Refractive Index 1.5060 to 1.5080
Quantity 250 g
Beilstein 17, 286
Specific Gravity 1.098
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 110.11
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Acetylfuran, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
EINECSNumber 214-757-1
RTECSNumber OB3870000
TSCA TSCA

RUO – Research Use Only