missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylfuran 98.0+%, TCI America™

Catalog No. A0091500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0091500G 500 g
A009125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0091500G Supplier TCI America Supplier No. A0091500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1192-62-7
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00003242
InChI Key IEMMBWWQXVXBEU-UHFFFAOYSA-N
Synonym 2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
PubChem CID 14505
ChEBI CHEBI:59983
IUPAC Name 1-(furan-2-yl)ethanone
SMILES CC(=O)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 173°C
Quantity 500 g
UN Number 2811
Formula Weight 110.11
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Acetylfuran
EINECSNumber (5)-3346
RTECSNumber OB3870000
TSCA Yes