missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-propylimidazolium iodide, 98%

Catalog No. AAH2768218
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2768218 50 g
AAH2768206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2768218 Supplier Thermo Scientific Chemicals Supplier No. H2768218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 119171-18-5 | C7H13IN2 | 252.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119171-18-5
Molecular Formula C7H13IN2
Molecular Weight (g/mol) 252.10
MDL Number MFCD08276385
InChI Key IVCMUVGRRDWTDK-UHFFFAOYSA-M
Synonym 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide
PubChem CID 12971008
IUPAC Name 1-methyl-3-propylimidazol-1-ium;iodide
SMILES [I-].CCCN1C=C[N+](C)=C1

Specifications

Density 1.54
Boiling Point 301°C to 304°C
Flash Point >110°C (230°F)
Quantity 50 g
Sensitivity Light sensitive
Formula Weight 252.09
Percent Purity 98%
Chemical Name or Material 1-Methyl-3-n-propylimidazolium iodide
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only