accessibility menu, dialog, popup

UserName

CAS RN 1191-67-9

IUPAC Name: decane-1,10-dithiol
Synonyms: decane-1,10-dithiol
Molecular Weight (g/mol): 206.41
Molecular Formula: C10H22S2
InChi Key: UOQACRNTVQWTFF-UHFFFAOYSA-N
SMILES: SCCCCCCCCCCS
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)