missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanedithiol, 95%

Catalog No. AAL1106414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1106414 25 g
AAL1106406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1106414 Supplier Thermo Scientific Chemicals Supplier No. L1106414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1191-67-9 | C10H22S2 | 206.41 g/mol

1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Decanedithiol is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Store under inert gas.

Chemical Identifiers

CAS 1191-67-9
Molecular Formula C10H22S2
Molecular Weight (g/mol) 206.41
MDL Number MFCD00022095
InChI Key UOQACRNTVQWTFF-UHFFFAOYSA-N
Synonym 1,10-decanedithiol, 1,10-dimercaptodecane, acmc-1bvry, 1,10-didecyl mercaptan, decamethylene dimercaptan
PubChem CID 14494
IUPAC Name decane-1,10-dithiol
SMILES SCCCCCCCCCCS

Specifications

Melting Point ∼17°C
Density 0.95
Boiling Point 171°C to 172°C (0.5 mmHg)
Odor Stench
Refractive Index 1.499
Quantity 25 g
UN Number UN3334
Beilstein 1738606
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 206.41
Percent Purity 95%
Chemical Name or Material 1,10-Decanedithiol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber 214-739-3
RTECSNumber HD8455000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only