missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanedithiol 98.0+%, TCI America™

Catalog No. D001525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D001525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D001525G Supplier TCI America Supplier No. D001525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used for chemical research.

Chemical Identifiers

CAS 1191-67-9
Molecular Formula C10H22S2
Molecular Weight (g/mol) 206.41
MDL Number MFCD00022095
InChI Key UOQACRNTVQWTFF-UHFFFAOYSA-N
Synonym 1,10-decanedithiol, 1,10-dimercaptodecane, acmc-1bvry, 1,10-didecyl mercaptan, decamethylene dimercaptan
PubChem CID 14494
IUPAC Name decane-1,10-dithiol
SMILES SCCCCCCCCCCS

Specifications

Color Yellow
Boiling Point 297°C
Quantity 25 g
Formula Weight 206.41
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,10-Decanedithiol
RTECSNumber HD8455000
TSCA No