accessibility menu, dialog, popup

UserName

HSSH

CAS RN 1191-62-4

IUPAC Name: octane-1,8-dithiol
Synonyms: octane-1,8-dithiol
Molecular Weight (g/mol): 178.35
Molecular Formula: C8H18S2
InChi Key: PGTWZHXOSWQKCY-UHFFFAOYSA-N
SMILES: SCCCCCCCCS
Boiling Point 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)