missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanedithiol, 99%

Catalog No. AC310620050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC310620050 5 g Glass bottle
AC310620010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310620050 Supplier Thermo Scientific Chemicals Supplier No. 310620050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1191-62-4 | C8H18S2 | 178.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1191-62-4
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00003574
InChI Key PGTWZHXOSWQKCY-UHFFFAOYSA-N
Synonym 1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474
PubChem CID 14493
IUPAC Name octane-1,8-dithiol
SMILES SCCCCCCCCS

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 269.0°C to 270.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5030 to 1.5050
Linear Formula HS(CH2)8SH
Quantity 5 g
Specific Gravity 0.96
Solubility Information Solubility in water: .. Other solubilities: soluble in most organic solvents
Formula Weight 178.36
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 8-Octanedithiol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia
EINECSNumber 214-738-8
RTECSNumber RG9610000
TSCA TSCA

RUO – Research Use Only