missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanedithiol 97.0+%, TCI America™

Catalog No. O03745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03745G Supplier TCI America Supplier No. O03745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical reagent intended for laboratory research purposes.

Chemical Identifiers

CAS 1191-62-4
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00003574
InChI Key PGTWZHXOSWQKCY-UHFFFAOYSA-N
Synonym 1,8-octanedithiol, 1,8-dimercaptooctane, octamethylene dimercaptan, 1,8-octamethylenedithiol, unii-kh3w3161hs, fema no. 3514, 1,8-octane dithiol, nanothinks tm thio8, acmc-209a1j, dsstox_cid_27474
PubChem CID 14493
IUPAC Name octane-1,8-dithiol
SMILES SCCCCCCCCS

Specifications

Melting Point 1°C
Color Colorless
Boiling Point 269°C
Quantity 5 g
Formula Weight 178.35
Percent Purity ≥97.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 1,8-Octanedithiol
RTECSNumber RG9610000
TSCA No