accessibility menu, dialog, popup

UserName

CAS RN 119-58-4

H3CNCH3OHNCH3H3C

CAS RN 119-58-4

IUPAC Name: bis[4-(dimethylamino)phenyl]methanol
Synonyms: bis[4-(dimethylamino)phenyl]methanol
Molecular Weight (g/mol): 270.38
Molecular Formula: C17H22N2O
InChi Key: YLZSIUVOIFJGQZ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(O)C1=CC=C(C=C1)N(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results