missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dimethylamino)benzhydrol 98.0+%, TCI America™

Catalog No. B048010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B048010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B048010G Supplier TCI America Supplier No. B048010G
Only null left

Chemical Identifiers

CAS 119-58-4
Molecular Formula C17H22N2O
Molecular Weight (g/mol) 270.38
MDL Number MFCD00008313
InChI Key YLZSIUVOIFJGQZ-UHFFFAOYSA-N
Synonym 4,4'-bis dimethylamino benzhydrol, michler's hydrol, bis 4-dimethylamino phenyl methanol, p,p'-michler's hydrol, tetramethyldiaminobenzhydrol, p,p'-bis dimethylamino benzhydrol, 4,4'-bis dimethylamino benzohydrol, 4,4'-bis dimethylamino diphenylmethanol, n,n'-tetramethyl-4,4'-diaminobenzohydrol
PubChem CID 92742
IUPAC Name bis[4-(dimethylamino)phenyl]methanol
SMILES CN(C)C1=CC=C(C=C1)C(O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 104°C
Quantity 10 g
Formula Weight 270.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(dimethylamino)benzhydrol
EINECSNumber (4)-0124
RTECSNumber DC7525000
TSCA Yes