accessibility menu, dialog, popup

UserName

CAS RN 1188-21-2

H3C(S)OHOHNOCH3CH3

CAS RN 1188-21-2

IUPAC Name: (2S)-2-acetamido-4-methylpentanoic acid
Synonyms: N-acetyl-leu
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: WXNXCEHXYPACJF-JLDDOWRYNA-N
SMILES: CC(C)C[C@H](NC(C)=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)