CAS RN 117752-04-2
IUPAC Name:
2-fluoro-6-methylbenzaldehyde
Synonyms:
2-fluoro-6-methylbenzaldehyde
Molecular Weight (g/mol):
138.14
Molecular Formula:
C8H7FO
InChi Key:
BGQBJWPZBBMKEI-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C(F)=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-6-methylbenzaldehyde, 97%
2-Fluoro-6-methylbenzaldehyde, 97%
CAS: 117752-04-2 Molecular Formula: C8H7FO Molecular Weight (g/mol): 138.14 MDL Number: MFCD09258962 InChI Key: BGQBJWPZBBMKEI-UHFFFAOYSA-N PubChem CID: 14170517 IUPAC Name: 2-fluoro-6-methylbenzaldehyde SMILES: CC1=C(C=O)C(F)=CC=C1