accessibility menu, dialog, popup

UserName

H3CNOFFFOH3C

CAS RN 116332-61-7

IUPAC Name: N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Synonyms: N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 233.19
Molecular Formula: C10H10F3NO2
InChi Key: YFDOMHRDNWBTMU-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results