missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methyl-4-(trifluoromethyl)benzamide 98.0+%, TCI America™

Catalog No. M27621G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27621G Supplier TCI America Supplier No. M27621G

Specifications

CAS 116332-61-7
Color Yellow
Molecular Formula C10H10F3NO2
Quantity 1 g
MDL Number MFCD08689795
InChI Key YFDOMHRDNWBTMU-UHFFFAOYSA-N
SMILES CN(C(=O)C1=CC=C(C=C1)C(F)(F)F)OC
IUPAC Name N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Molecular Weight (g/mol) 233.19
PubChem CID 14069348
Formula Weight 233.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methyl-4-(trifluoromethyl)benzamide
Show More Show Less