CAS RN 1156-50-9
IUPAC Name:
1-nitro-4-(4-nitrobenzenesulfonyl)benzene
Synonyms:
1-nitro-4-(4-nitrobenzenesulfonyl)benzene
Molecular Weight (g/mol):
308.26
Molecular Formula:
C12H8N2O6S
InChi Key:
BVHNGWRPAFKGFP-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O