missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) Sulfone 98.0+%, TCI America™

Catalog No. D19275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D19275G Supplier TCI America Supplier No. D19275G

Specifications

CAS 1156-50-9
Color Yellow
Molecular Formula C12H8N2O6S
Quantity 5 g
MDL Number MFCD00010447
Synonym bis 4-nitrophenyl sulfone, 4-nitrophenyl sulfone, 4,4'-dinitrodiphenyl sulfone, bis 4-nitrophenyl sulphone, bis p-nitrophenyl sulfone, 4,4'-dinitrodiphenylsulfone, 1,1'-sulfonylbis 4-nitrobenzene, sulfone, bis p-nitrophenyl, benzene, 1,1'-sulfonylbis 4-nitro-9ci
InChI Key BVHNGWRPAFKGFP-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]
IUPAC Name 1-nitro-4-(4-nitrophenyl)sulfonylbenzene
Molecular Weight (g/mol) 308.264
PubChem CID 70871
Formula Weight 308.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrophenyl) Sulfone
Show More Show Less