accessibility menu, dialog, popup

UserName

NNN

CAS RN 1148-79-4

IUPAC Name: 6-(pyridin-2-yl)-2,2'-bipyridine
Synonyms: terpyridine α,α',α''-tripyridyl tripyridyl α,α',α''-terpyridine 2,2',2''-tripyridyl terpy 2,2',2''-terpyridyl 2,2',2''-tripyridine 2,2',2''-terpyridine tripyridine
Molecular Weight (g/mol): 233.27
Molecular Formula: C15H11N3
InChi Key: DRGAZIDRYFYHIJ-UHFFFAOYSA-N
SMILES: C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)