missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2″-Terpyridine 98.0+%, TCI America™

Catalog No. T0024200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0024200MG 200 mg
T00241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0024200MG Supplier TCI America Supplier No. T0024200MG

Chemical Identifiers

CAS 1148-79-4
Molecular Formula C15H11N3
Molecular Weight (g/mol) 233.27
MDL Number MFCD00006213
InChI Key DRGAZIDRYFYHIJ-UHFFFAOYSA-N
Synonym 2,2':6',2-terpyridine, tripyridyl, tripyridine, 2,6-bis 2-pyridyl pyridine, terpy, terpyridine, 2,2',2-terpyridine, 2,2',2-terpyridyl, 2,2',2-tripyridyl, 2,2',2-tripyridine
PubChem CID 70848
ChEBI CHEBI:245199
IUPAC Name 6-(pyridin-2-yl)-2,2'-bipyridine
SMILES C1=CC=C(N=C1)C1=CC=CC(=N1)C1=CC=CC=N1

Specifications

Melting Point 92°C
Color Green-White
Boiling Point 370°C
Quantity 200 mg
UN Number 2811
Formula Weight 233.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′:6′,2′′-Terpyridine