accessibility menu, dialog, popup

UserName

HOOHNONHO

CAS RN 1145-32-0

IUPAC Name: 2-[2-(phenylformamido)acetamido]acetic acid
Synonyms: [2-(phenylformamido)acetamido]acetic acid
Molecular Weight (g/mol): 236.23
Molecular Formula: C11H12N2O4
InChi Key: FWYPSSZPXQVFRB-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)CNC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results