missing translation for 'onlineSavingsMsg'
Learn More

Benzoylglycylglycine 98.0+%, TCI America™

Catalog No. B02031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B02031G Supplier TCI America Supplier No. B02031G

Specifications

Purity ≥98.0% (HPLC,T)
CAS 1145-32-0
Color White-Yellow
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 236.23
Product Type Benzoylglycylglycine
Quantity 1 g