accessibility menu, dialog, popup

UserName

FNI

CAS RN 113975-22-7

IUPAC Name: 2-fluoro-3-iodopyridine
Synonyms: 2-fluoro-3-iodopyridine
Molecular Weight (g/mol): 222.99
Molecular Formula: C5H3FIN
InChi Key: WCDCAXVNBOLWNO-UHFFFAOYSA-N
SMILES: FC1=NC=CC=C1I
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)