missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-iodopyridine, 97%, Thermo Scientific™

Catalog No. AAH5014603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5014603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5014603 Supplier Thermo Scientific Chemicals Supplier No. H5014603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 113975-22-7
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD03095287
InChI Key WCDCAXVNBOLWNO-UHFFFAOYSA-N
Synonym 2-fluoro-3-iodo-pyridine, pyridine, 2-fluoro-3-iodo, zlchem 409, 2-fluoro-3 iodopyridine, 2-fluoro-3-iodo-pyridin, 2-fluoro-3-iodopyridine, 2-fluoro-3-iodopyridine;, 2-fluoro-3-iodo pyridine, acmc-1c73w, ksc174s0n
PubChem CID 7009501
IUPAC Name 2-fluoro-3-iodopyridine
SMILES C1=CC(=C(N=C1)F)I

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 222.99
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-iodopyridine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures