missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-iodopyridine 98.0+%, TCI America™

Catalog No. F07335G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07335G 5 g
F07331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07335G Supplier TCI America Supplier No. F07335G

Chemical Identifiers

CAS 113975-22-7
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD03095287
InChI Key WCDCAXVNBOLWNO-UHFFFAOYSA-N
Synonym 2-fluoro-3-iodo-pyridine, pyridine, 2-fluoro-3-iodo, zlchem 409, 2-fluoro-3 iodopyridine, 2-fluoro-3-iodo-pyridin, 2-fluoro-3-iodopyridine, 2-fluoro-3-iodopyridine;, 2-fluoro-3-iodo pyridine, acmc-1c73w, ksc174s0n
PubChem CID 7009501
IUPAC Name 2-fluoro-3-iodopyridine
SMILES C1=CC(=C(N=C1)F)I

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 99°C
Quantity 5 g
Formula Weight 222.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-3-iodopyridine