CAS RN 1133798-41-0
IUPAC Name:
tris(3,4-dichlorophenyl)-1,3,5,2,4,6-trioxatriborinane
Synonyms:
tris(3,4-dichlorophenyl)-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol):
518.4
Molecular Formula:
C18H9B3Cl6O3
InChi Key:
ACMLOUSDQIZBAK-UHFFFAOYSA-N
SMILES:
ClC1=C(Cl)C=C(C=C1)B1OB(OB(O1)C1=CC(Cl)=C(Cl)C=C1)C1=CC(Cl)=C(Cl)C=C1
1
–
1
of
1
results
1
2,4,6-Tris(3,4-dichlorophenyl)boroxin 98.0+%, TCI America™
CAS: 1133798-41-0 Molecular Formula: C18H9B3Cl6O3 Molecular Weight (g/mol): 518.397 InChI Key: ACMLOUSDQIZBAK-UHFFFAOYSA-N Synonym: Tris(3,4-dichlorophenyl)cycloboroxane, 3,4-Dichlorophenylboronic Anhydride PubChem CID: 44629844 IUPAC Name: 2,4,6-tris(3,4-dichlorophenyl)-1,3,5,2,4,6-trioxatriborinane SMILES: B1(OB(OB(O1)C2=CC(=C(C=C2)Cl)Cl)C3=CC(=C(C=C3)Cl)Cl)C4=CC(=C(C=C4)Cl)Cl