accessibility menu, dialog, popup

UserName

OHONH2F

CAS RN 1132-68-9

IUPAC Name: 2-amino-3-(4-fluorophenyl)propanoic acid
Synonyms: DL-P-fluorophenylalanine D,L-fluorophenylalanine 4-fluoro-dl-para-phenylalanine alnasid D,L-p-fluorophenylalanine
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: XWHHYOYVRVGJJY-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(F)C=C1)C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)