missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-L-phenylalanine, 98+%

Catalog No. AAL19934MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19934MD 250 mg
AAL1993403 1 g
AAL1993406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19934MD Supplier Thermo Scientific Chemicals Supplier No. L19934MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1132-68-9 | C9H10FNO2 | 183.182 g/mol

4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

Solubility
Soluble in water and 0.5M HCl (50 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1132-68-9
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00063064
InChI Key XWHHYOYVRVGJJY-QMMMGPOBSA-N
Synonym 4-fluoro-l-phenylalanine, p-fluoro-l-phenylalanine, s-4-fluorophenylalanine, l-4-fluorophenylalanine, h-phe 4-f-oh, s-2-amino-3-4-fluorophenyl propanoic acid, unii-e2k2vdk6kk, l-phenylalanine, 4-fluoro, s---p-fluorophenylalanine, 2s-2-amino-3-4-fluorophenyl propanoic acid
PubChem CID 716312
ChEBI CHEBI:44909
IUPAC Name (2S)-2-amino-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)F

Specifications

Melting Point ∽240°C (decomposition)
Beilstein 2416148
Solubility Information Soluble in water and 0.5M HCl (50mg/ml).
Quantity 250 mg
Formula Weight 183.18
Optical Rotation −25° (c=1 in Water)
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-L-phenylalanine
RTECSNumber DW1765000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only