missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-L-phenylalanine 98.0+%, TCI America™

Catalog No. F0274100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0274100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0274100MG Supplier TCI America Supplier No. F0274100MG

Chemical Identifiers

CAS 1132-68-9
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00063064
InChI Key XWHHYOYVRVGJJY-QMMMGPOBSA-N
Synonym 4-fluoro-l-phenylalanine, p-fluoro-l-phenylalanine, s-4-fluorophenylalanine, l-4-fluorophenylalanine, h-phe 4-f-oh, s-2-amino-3-4-fluorophenyl propanoic acid, unii-e2k2vdk6kk, l-phenylalanine, 4-fluoro, s---p-fluorophenylalanine, 2s-2-amino-3-4-fluorophenyl propanoic acid
PubChem CID 716312
ChEBI CHEBI:44909
IUPAC Name (2S)-2-amino-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)F

Specifications

Color White
Quantity 100 mg
Formula Weight 183.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-L-phenylalanine